Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD7
Entrez Gene ID
79955
Gene Name
PDZ domain containing 7
Gene Aliases
PDZK7
Chromosome Location
Chr.10: 101007683 - 101031157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195263.1 NP_001182192.1 1 - 2 85 75
NM_024895.4 NP_079171.1 1 - 2 85 75
XM_005270165.2 XP_005270222.1 1 - 2 60 75
XM_011540178.2 XP_011538480.1 1 - 2 60 75
XM_011540179.2 XP_011538481.1 1 - 2 60 75
XM_011540181.2 XP_011538483.1 1 - 2 60 75
XM_011540182.2 XP_011538484.1 1 - 2 60 75
XM_011540183.2 XP_011538485.1 1 - 2 60 75
XM_017016667.1 XP_016872156.1 1 - 2 60 75
XM_017016668.1 XP_016872157.1 1 - 2 60 75
GenBank mRNA AK026862.1 - 1 - 2 50 75
AK225136.1 - 1 - 2 57 75
DA197280.1 - 1 - 2 86 75
FJ617449.1 - 1 - 2 49 75

View Full Product Details