Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DERA
Entrez Gene ID
51071
Gene Name
deoxyribose-phosphate aldolase
Gene Aliases
CGI-26, DEOC
Chromosome Location
Chr.12: 15911172 - 16037381 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300779.1 NP_001287708.1 2 - 3 345 115
NM_015954.3 NP_057038.2 2 - 3 345 115
GenBank mRNA AF132960.1 - 2 - 3 206 115
AK024614.1 - 2 - 3 161 115
AK222541.1 - 2 - 3 161 115
AK309182.1 - 2 - 3 261 115
BC041328.1 - 2 - 3 345 115
BC056234.1 - 2 - 3 187 115
BU928832.1 - 2 - 3 151 115

View Full Product Details