The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300779.1 | NP_001287708.1 | 6 - 7 | 824 | 68 |
NM_015954.3 | NP_057038.2 | 7 - 8 | 953 | 68 | |
GenBank mRNA | AF132960.1 | - | 7 - 8 | 814 | 68 |
AK309182.1 | - | 7 - 8 | 869 | 68 | |
BC041328.1 | - | 7 - 8 | 953 | 68 | |
BC056234.1 | - | 7 - 8 | 795 | 68 |