The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006717519.3 | XP_006717582.1 | 11 - 12 | 3347 | 130 |
XM_017016852.1 | XP_016872341.1 | 10 - 11 | 3329 | 130 |