Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP5K1P1
Entrez Gene ID
206426
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 pseudogene 1
Gene Aliases
-
Chromosome Location
Chr.6: 7986102 - 7990344 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027712.1 1 - 1 2218 124
GenBank mRNA BC028580.2 - 1 - 1 2217 124

View Full Product Details