The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020129.2 | NP_064514.1 | 2 - 3 | 317 | 121 |
NM_203471.1 | NP_982297.1 | 3 - 4 | 622 | 121 | |
GenBank mRNA | AF267852.1 | - | 2 - 3 | 317 | 121 |
AF367430.1 | - | 2 - 3 | 317 | 121 | |
AF367431.1 | - | 2 - 3 | 317 | 121 | |
AF367432.1 | - | 2 - 3 | 317 | 121 | |
AK312040.1 | - | 2 - 3 | 156 | 121 | |
BC022257.1 | - | 2 - 3 | 173 | 121 | |
BG434228.1 | - | 3 - 4 | 448 | 121 | |
FJ613341.1 | - | 2 - 3 | 121 | 121 | |
FJ613342.1 | - | 2 - 3 | 121 | 121 |