Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB3IL1
Entrez Gene ID
5866
Gene Name
RAB3A interacting protein like 1
Gene Aliases
GRAB
Chromosome Location
Chr.11: 61897296 - 61946312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271686.1 NP_001258615.1 2 - 3 477 73
NM_013401.3 NP_037533.2 2 - 3 501 73
XM_005274140.4 XP_005274197.1 2 - 3 466 73
XM_006718647.3 XP_006718710.1 2 - 3 468 73
XM_011545196.2 XP_011543498.1 2 - 3 472 73
XM_011545197.2 XP_011543499.1 2 - 3 788 73
XM_011545198.2 XP_011543500.1 2 - 3 496 73
XM_011545199.2 XP_011543501.1 2 - 3 474 73
XM_011545201.2 XP_011543503.1 2 - 3 479 73
GenBank mRNA AF084557.1 - 1 - 2 55 73
BC022239.1 - 2 - 3 417 73
BC051820.1 - 2 - 3 477 73
BQ946081.1 - 2 - 3 443 73
DR004757.1 - 2 - 3 503 73

View Full Product Details