Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB3IL1
Entrez Gene ID
5866
Gene Name
RAB3A interacting protein like 1
Gene Aliases
GRAB
Chromosome Location
Chr.11: 61897296 - 61946312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013401.3 NP_037533.2 4 - 5 684 103
XM_005274140.4 XP_005274197.1 4 - 5 649 103
XM_006718647.3 XP_006718710.1 4 - 5 651 103
XM_011545196.2 XP_011543498.1 4 - 5 655 103
XM_011545197.2 XP_011543499.1 4 - 5 971 103
XM_011545198.2 XP_011543500.1 4 - 5 679 103
GenBank mRNA AF084557.1 - 3 - 4 238 103
BC022239.1 - 4 - 5 600 103
BQ946081.1 - 4 - 5 626 104

View Full Product Details