Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB3IL1
Entrez Gene ID
5866
Gene Name
RAB3A interacting protein like 1
Gene Aliases
GRAB
Chromosome Location
Chr.11: 61897296 - 61946312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271686.1 NP_001258615.1 5 - 6 783 52
NM_013401.3 NP_037533.2 6 - 7 1026 52
XM_005274140.4 XP_005274197.1 6 - 7 991 52
XM_006718647.3 XP_006718710.1 6 - 7 993 52
XM_011545196.2 XP_011543498.1 6 - 7 997 52
XM_011545197.2 XP_011543499.1 6 - 7 1313 52
XM_011545198.2 XP_011543500.1 6 - 7 1021 52
XM_011545199.2 XP_011543501.1 5 - 6 780 52
XM_011545200.2 XP_011543502.1 3 - 4 432 52
GenBank mRNA AF084557.1 - 5 - 6 580 52
BC022239.1 - 6 - 7 942 52
BC051820.1 - 5 - 6 783 52

View Full Product Details