The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322464.1 | NP_001309393.1 | 4 - 5 | 1257 | 67 |
NM_030645.1 | NP_085148.1 | 4 - 5 | 927 | 67 | |
XM_017002404.1 | XP_016857893.1 | 3 - 4 | 1130 | 67 | |
XM_017002405.1 | XP_016857894.1 | 3 - 4 | 909 | 67 | |
XM_017002406.1 | XP_016857895.1 | 5 - 6 | 883 | 67 | |
XM_017002407.1 | XP_016857896.1 | 3 - 4 | 815 | 67 | |
GenBank mRNA | AB051507.1 | - | 3 - 4 | 634 | 67 |
AK056345.1 | - | 1 - 2 | 112 | 67 | |
AK056382.1 | - | 4 - 5 | 927 | 67 | |
AK125138.1 | - | 4 - 5 | 1604 | 67 | |
AK298698.1 | - | 3 - 4 | 281 | 67 | |
AL136569.1 | - | 3 - 4 | 514 | 67 | |
AY358453.1 | - | 4 - 5 | 849 | 67 | |
BC010871.1 | - | 1 - 2 | 136 | 67 | |
BC017254.2 | - | 3 - 4 | 416 | 67 |