The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322462.1 | NP_001309391.1 | 8 - 9 | 1935 | 65 |
NM_001322463.1 | NP_001309392.1 | 6 - 7 | 1310 | 65 | |
NM_001322464.1 | NP_001309393.1 | 6 - 7 | 1677 | 65 | |
NM_030645.1 | NP_085148.1 | 6 - 7 | 1264 | 65 | |
XM_017002404.1 | XP_016857893.1 | 5 - 6 | 1467 | 65 | |
XM_017002405.1 | XP_016857894.1 | 5 - 6 | 1246 | 65 | |
XM_017002406.1 | XP_016857895.1 | 7 - 8 | 1220 | 65 | |
XM_017002407.1 | XP_016857896.1 | 5 - 6 | 1152 | 65 | |
GenBank mRNA | AB051507.1 | - | 5 - 6 | 971 | 65 |
AK056345.1 | - | 3 - 4 | 449 | 65 | |
AK056382.1 | - | 6 - 7 | 1264 | 65 | |
AK091164.1 | - | 1 - 2 | 1186 | 65 | |
AK125138.1 | - | 6 - 7 | 1941 | 65 | |
AK125837.1 | - | 8 - 9 | 2204 | 65 | |
AK298698.1 | - | 5 - 6 | 618 | 65 | |
AK299713.1 | - | 2 - 3 | 628 | 65 | |
AL136569.1 | - | 5 - 6 | 851 | 65 | |
AY358453.1 | - | 6 - 7 | 1186 | 65 | |
BC010871.1 | - | 3 - 4 | 473 | 65 | |
BC017254.2 | - | 5 - 6 | 753 | 65 |