The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126340.2 | NP_001119812.1 | 1 - 2 | 129 | 76 |
NM_001271819.1 | NP_001258748.1 | 1 - 2 | 129 | 76 | |
GenBank mRNA | AK300574.1 | - | 1 - 2 | 76 | |
DA770788.1 | - | 1 - 2 | 124 | 76 | |
HY064081.1 | - | 1 - 2 | 130 | 76 |