The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127603.1 | NP_001121075.1 | 4 - 5 | 412 | 103 |
NM_017881.2 | NP_060351.1 | 4 - 5 | 412 | 103 | |
NR_023352.1 | 5 - 6 | 669 | 103 | ||
XM_006717162.1 | XP_006717225.1 | 5 - 6 | 697 | 103 | |
XM_006717163.2 | XP_006717226.1 | 5 - 6 | 732 | 103 | |
XM_011518817.1 | XP_011517119.1 | 4 - 5 | 522 | 103 | |
XM_011518818.1 | XP_011517120.1 | 4 - 5 | 1044 | 103 | |
XM_011518819.2 | XP_011517121.1 | 4 - 5 | 1044 | 103 | |
XM_017014868.1 | XP_016870357.1 | 6 - 7 | 1018 | 103 | |
XM_017014869.1 | XP_016870358.1 | 5 - 6 | 1325 | 103 | |
XM_017014870.1 | XP_016870359.1 | 4 - 5 | 1068 | 103 | |
XM_017014871.1 | XP_016870360.1 | 5 - 6 | 697 | 103 | |
XM_017014872.1 | XP_016870361.1 | 6 - 7 | 1018 | 103 | |
XM_017014873.1 | XP_016870362.1 | 6 - 7 | 989 | 103 | |
XM_017014874.1 | XP_016870363.1 | 5 - 6 | 732 | 103 | |
GenBank mRNA | AK000566.1 | - | 4 - 5 | 377 | 103 |
AK023413.1 | - | 4 - 5 | 404 | 103 | |
AK097144.1 | - | 5 - 6 | 642 | 103 | |
AY611480.1 | - | 3 - 4 | 167 | 103 | |
BC001366.2 | - | 4 - 5 | 320 | 103 | |
BC036804.1 | - | 4 - 5 | 412 | 103 |