Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
Chromosome Location
Chr.7: 978485 - 1138291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032350.5 NP_115726.1 1 - 2 67 123
XM_011515581.2 XP_011513883.1 1 - 2 101 123
XM_017012720.1 XP_016868209.1 1 - 2 104
XM_017012722.1 XP_016868211.1 1 - 2 100 123
GenBank mRNA BC006224.2 - 1 - 2 67 123

View Full Product Details