The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282062.1 | NP_001268991.1 | 4 - 5 | 570 | 64 |
NM_001282063.1 | NP_001268992.1 | 4 - 5 | 532 | 64 | |
NM_001282068.1 | NP_001268997.1 | 3 - 4 | 497 | 64 | |
NM_003586.2 | NP_003577.2 | 3 - 4 | 531 | 64 | |
NR_104089.1 | 3 - 4 | 322 | 64 | ||
NR_104090.1 | 3 - 4 | 322 | 64 | ||
XM_011545975.1 | XP_011544277.1 | 5 - 6 | 956 | 64 | |
XM_017023776.1 | XP_016879265.1 | 7 - 8 | 1174 | 64 | |
XM_017023777.1 | XP_016879266.1 | 4 - 5 | 1111 | 64 | |
XM_017023778.1 | XP_016879267.1 | 3 - 4 | 585 | 64 | |
GenBank mRNA | AK293651.1 | - | 3 - 4 | 271 | 64 |
BC041769.1 | - | 4 - 5 | 533 | 64 | |
BC055284.1 | - | 3 - 4 | 531 | 64 | |
BC063436.1 | - | 4 - 5 | 496 | 64 | |
BI757951.1 | - | 5 - 6 | 442 | 64 | |
BX460259.2 | - | 3 - 4 | 251 | 64 | |
DC399462.1 | - | 5 - 6 | 474 | 64 |