The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000353.2 | NP_000344.1 | 8 - 9 | 1013 | 79 |
GenBank mRNA | AK313380.1 | - | 8 - 9 | 1014 | 79 |
BC130534.1 | - | 8 - 9 | 984 | 79 | |
X52520.1 | - | 8 - 9 | 1010 | 79 | |
X55675.1 | - | 8 - 9 | 964 | 79 |