The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005474.2 | NP_001005474.1 | 4 - 5 | 447 | 100 |
NM_031419.3 | NP_113607.1 | 3 - 4 | 580 | 100 | |
GenBank mRNA | AB037925.1 | - | 3 - 4 | 513 | 100 |
AF548362.1 | - | 4 - 5 | 444 | 100 | |
AK054787.1 | - | 4 - 5 | 316 | 100 | |
AK091782.1 | - | 3 - 4 | 580 | 100 | |
BC014587.2 | - | - | 227 | 100 | |
BC060800.1 | - | 3 - 4 | 580 | 100 | |
DA541852.1 | - | 4 - 5 | 447 | 100 | |
DQ224339.1 | - | 3 - 4 | 369 | 100 |