Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFKBIZ
Entrez Gene ID
64332
Gene Name
NFKB inhibitor zeta
Gene Aliases
IKBZ, INAP, MAIL
Chromosome Location
Chr.3: 101827990 - 101861025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005474.2 NP_001005474.1 10 - 11 1813 68
NM_031419.3 NP_113607.1 9 - 10 1946 68
GenBank mRNA AB037925.1 - 9 - 10 1879 68
AF548362.1 - 10 - 11 1810 68
AK054787.1 - 10 - 11 1682 68
AK091782.1 - 10 - 11 1580 68
BC014587.2 - - 1862 68
BC060800.1 - 9 - 10 1946 68
DQ224339.1 - 9 - 10 1735 68

View Full Product Details