The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080660.3 | NP_542391.2 | 1 - 2 | 371 | 74 |
XM_006716176.3 | XP_006716239.1 | 1 - 2 | 814 | 74 | |
XM_011516688.2 | XP_011514990.1 | 1 - 2 | 812 | 74 | |
GenBank mRNA | BC008842.2 | - | 1 - 2 | 371 | 74 |