Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTNNA1
Entrez Gene ID
1495
Gene Name
catenin alpha 1
Gene Aliases
CAP102, MDPT2
Chromosome Location
Chr.5: 138753386 - 138935034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290307.2 NP_001277236.1 2 - 3 233 66
NM_001290310.2 NP_001277239.1 2 - 3 233 66
NM_001323982.1 NP_001310911.1 3 - 4 703 66
NM_001323983.1 NP_001310912.1 2 - 3 385 66
NM_001323984.1 NP_001310913.1 3 - 4 251 66
NM_001323985.1 NP_001310914.1 2 - 3 233 66
NM_001323986.1 NP_001310915.1 2 - 3 233 66
NM_001903.4 NP_001894.2 2 - 3 233 66
GenBank mRNA AK094151.1 - 2 - 3 172 66
AK296708.1 - 2 - 3 204 66
AK304267.1 - 2 - 3 173 66
BC000385.2 - 2 - 3 171 66
D13866.1 - 2 - 3 172 66
D14705.1 - 2 - 3 197 66
DC332608.1 - 2 - 3 233 66
DC369291.1 - 2 - 3 223 66
L23805.1 - 1 - 2 110 66
U03100.1 - 1 - 2 110 66

View Full Product Details