Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location
Chr.8: 61500556 - 61714640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164750.1 NP_001158222.1 14 - 15 1048 128
NM_004318.3 NP_004309.2 14 - 15 1243 128
XM_005251235.2 XP_005251292.1 15 - 16 1257 128
XM_005251236.2 XP_005251293.1 15 - 16 1254 128
XM_005251239.2 XP_005251296.1 14 - 15 1200 128
XM_005251240.1 XP_005251297.1 15 - 16 1035 128
XM_005251242.2 XP_005251299.1 13 - 14 1155 128
XM_005251243.2 XP_005251300.1 14 - 15 1128 128
XM_005251244.1 XP_005251301.1 14 - 15 975 128
XM_005251246.2 XP_005251303.1 13 - 14 1083 128
XM_005251247.2 XP_005251304.1 13 - 14 1071 128
XM_005251248.1 XP_005251305.1 13 - 14 841 128
XM_005251250.2 XP_005251307.1 12 - 13 1026 128
XM_017013419.1 XP_016868908.1 15 - 16 1257 128
XM_017013420.1 XP_016868909.1 15 - 16 1254 128
XM_017013421.1 XP_016868910.1 14 - 15 1215 128
XM_017013422.1 XP_016868911.1 14 - 15 1212 128
XM_017013424.1 XP_016868913.1 14 - 15 1209 128
XM_017013425.1 XP_016868914.1 14 - 15 1200 128
XM_017013426.1 XP_016868915.1 15 - 16 1035 128
XM_017013427.1 XP_016868916.1 15 - 16 1032 128
XM_017013428.1 XP_016868917.1 13 - 14 1155 128
XM_017013429.1 XP_016868918.1 14 - 15 1209 128
XM_017013430.1 XP_016868919.1 14 - 15 1128 128
XM_017013431.1 XP_016868920.1 14 - 15 898 128
XM_017013432.1 XP_016868921.1 14 - 15 1125 128
XM_017013433.1 XP_016868922.1 14 - 15 895 128
XM_017013434.1 XP_016868923.1 14 - 15 1197 128
XM_017013435.1 XP_016868924.1 14 - 15 975 128
XM_017013437.1 XP_016868926.1 13 - 14 1083 128
XM_017013438.1 XP_016868927.1 14 - 15 1125 128
XM_017013439.1 XP_016868928.1 14 - 15 895 128
XM_017013440.1 XP_016868929.1 14 - 15 906 128
XM_017013441.1 XP_016868930.1 13 - 14 1080 128
XM_017013442.1 XP_016868931.1 13 - 14 1068 128
XM_017013443.1 XP_016868932.1 12 - 13 1023 128
XM_017013444.1 XP_016868933.1 13 - 14 769 128
GenBank mRNA AK304314.1 - 14 - 15 1004 128
S83325.1 - 14 - 15 985 128
U03109.1 - 14 - 15 1051 128

View Full Product Details