The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308350.1 | NP_001295279.1 | 5 - 6 | 1312 | 109 |
NM_001322294.1 | NP_001309223.1 | 5 - 6 | 1312 | 109 | |
NM_001322297.1 | NP_001309226.1 | 4 - 5 | 1190 | 109 | |
NM_001322299.1 | NP_001309228.1 | 5 - 6 | 1312 | 109 | |
NM_017699.2 | NP_060169.2 | 5 - 6 | 1312 | 109 | |
NR_136280.1 | 6 - 7 | 1392 | 109 | ||
NR_136282.1 | 5 - 6 | 1312 | 109 | ||
NR_136283.1 | 5 - 6 | 1312 | 109 | ||
XM_011512939.2 | XP_011511241.1 | 5 - 6 | 1272 | 109 | |
XM_011512941.2 | XP_011511243.1 | 5 - 6 | 1272 | 109 | |
XM_017006666.1 | XP_016862155.1 | 2 - 3 | 180 | 109 | |
GenBank mRNA | BC117222.1 | - | 5 - 6 | 700 | 109 |
BC143776.1 | - | 5 - 6 | 700 | 109 | |
BC143778.1 | - | 5 - 6 | 700 | 109 |