The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004843.3 | NP_004834.1 | 4 - 5 | 666 | 110 |
GenBank mRNA | AF053004.1 | - | 4 - 5 | 671 | 110 |
AF106912.1 | - | 4 - 5 | 658 | 110 | |
AF265242.1 | - | 4 - 5 | 644 | 110 | |
AY358876.1 | - | 4 - 5 | 644 | 110 | |
BC028003.1 | - | 4 - 5 | 589 | 110 |