Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
Chromosome Location
Chr.9: 123379654 - 123930158 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020946.1 NP_065997.1 16 - 17 1460 95
NM_024820.2 NP_079096.2 16 - 17 1460 95
XM_005252109.2 XP_005252166.1 16 - 17 1309 95
XM_005252111.4 XP_005252168.1 16 - 17 2276 95
XM_005252113.4 XP_005252170.1 16 - 17 2265 95
XM_006717195.3 XP_006717258.1 16 - 17 2277 95
XM_011518882.2 XP_011517184.1 16 - 17 2279 95
XM_011518883.2 XP_011517185.1 16 - 17 2280 95
XM_011518884.2 XP_011517186.1 15 - 16 2230 95
XM_011518885.2 XP_011517187.1 15 - 16 1972 95
XM_011518886.2 XP_011517188.1 16 - 17 2277 95
XM_011518887.2 XP_011517189.1 16 - 17 2268 95
XM_017014947.1 XP_016870436.1 15 - 16 2235 95
XM_017014948.1 XP_016870437.1 16 - 17 1309 95
XM_017014949.1 XP_016870438.1 15 - 16 2154 95
XM_017014950.1 XP_016870439.1 15 - 16 2153 95
XM_017014951.1 XP_016870440.1 15 - 16 1253 95
XM_017014952.1 XP_016870441.1 14 - 15 1390 95
GenBank mRNA AK024782.1 - 16 - 17 1448 95
AK074151.1 - 11 - 12 888 95
AK225104.1 - 16 - 17 1448 95
AK295710.1 - 14 - 15 1321 95
AK299867.1 - 7 - 8 522 95
BC009616.2 - 15 - 16 1428 95
BC028061.1 - 16 - 17 1250 95
BC039703.1 - 16 - 17 1313 95

View Full Product Details