The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256054.2 | NP_001242983.1 | 8 - 9 | 1289 | 88 |
NM_018325.4 | NP_060795.1 | 8 - 9 | 1194 | 88 | |
GenBank mRNA | AK001971.1 | - | 1 - 2 | 136 | 88 |
AK291425.1 | - | 8 - 9 | 1184 | 88 | |
AL832467.1 | - | 7 - 8 | 2768 | 88 | |
BC068445.1 | - | 7 - 8 | 1143 | 88 | |
JN681271.1 | - | 8 - 9 | 1267 | 88 |