Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEK
Entrez Gene ID
7010
Gene Name
TEK receptor tyrosine kinase
Gene Aliases
CD202B, TIE-2, TIE2, VMCM, VMCM1
Chromosome Location
Chr.9: 27109141 - 27230178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000459.4 NP_000450.2 21 - 22 3640 72
NM_001290077.1 NP_001277006.1 20 - 21 3511 72
NM_001290078.1 NP_001277007.1 19 - 20 3196 72
XM_005251561.1 XP_005251618.1 21 - 22 3304 72
XM_005251563.1 XP_005251620.1 20 - 21 3175 72
GenBank mRNA AB208796.1 - 21 - 22 3318 72
AK291775.1 - 21 - 22 3328 72
AK294887.1 - 19 - 20 3197 72
AK295043.1 - 20 - 21 3237 72
BC035514.1 - 21 - 22 3315 72
L06139.1 - 21 - 22 3338 72

View Full Product Details