The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000459.4 | NP_000450.2 | 6 - 7 | 1355 | 77 |
XM_005251561.1 | XP_005251618.1 | 6 - 7 | 1022 | 77 | |
GenBank mRNA | AB208796.1 | - | 6 - 7 | 1033 | 77 |
AK291775.1 | - | 6 - 7 | 1043 | 77 | |
BC035514.1 | - | 6 - 7 | 1030 | 77 | |
EU826591.1 | - | 6 - 7 | 905 | 77 | |
L06139.1 | - | 6 - 7 | 1053 | 77 |