The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290325.1 | NP_001277254.1 | 2 - 3 | 1352 | 88 |
NM_001290326.1 | NP_001277255.1 | 2 - 3 | 1352 | 88 | |
NM_032557.6 | NP_115946.2 | 2 - 3 | 1352 | 88 | |
XM_011532360.2 | XP_011530662.1 | 2 - 3 | 840 | 88 | |
GenBank mRNA | AK093754.1 | - | 2 - 3 | 575 | 88 |
AK126943.1 | - | 3 - 4 | 1180 | 88 | |
AL833976.1 | - | 2 - 3 | 1324 | 88 | |
BC039115.1 | - | 2 - 3 | 1352 | 88 | |
BC068975.1 | - | 2 - 3 | 1299 | 88 |