The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020946.1 | NP_065997.1 | 20 - 21 | 1809 | 75 |
XM_005252111.4 | XP_005252168.1 | 20 - 21 | 2625 | 75 | |
XM_011518886.2 | XP_011517188.1 | 21 - 22 | 2680 | 75 | |
GenBank mRNA | AK074151.1 | - | 15 - 16 | 1237 | 75 |