The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317851.1 | NP_001304780.1 | 10 - 11 | 1278 | 102 |
NM_016173.4 | NP_057257.1 | 10 - 11 | 1579 | 102 | |
XM_005265218.3 | XP_005265275.1 | 8 - 9 | 1423 | 102 | |
GenBank mRNA | AF131220.1 | - | 9 - 10 | 1165 | 102 |
AF172244.1 | - | 10 - 11 | 1179 | 102 | |
AK025973.1 | - | 10 - 11 | 1278 | 102 | |
BC000781.2 | - | 9 - 10 | 1147 | 102 | |
BX537562.1 | - | 9 - 10 | 1391 | 102 |