The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317851.1 | NP_001304780.1 | 1 - 2 | 122 | 89 |
XM_011533808.1 | XP_011532110.1 | 1 - 2 | 94 | 89 | |
GenBank mRNA | AK025973.1 | - | 1 - 2 | 122 | 89 |