The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206957.1 | NP_001193886.1 | 2 - 3 | 489 | 67 |
NM_007182.4 | NP_009113.3 | 3 - 4 | 596 | 67 | |
NM_170712.2 | NP_733830.1 | 3 - 4 | 306 | 67 | |
NM_170713.2 | NP_733831.1 | 2 - 3 | 400 | 67 | |
NM_170714.1 | NP_733832.1 | 3 - 4 | 608 | 67 | |
XM_011533315.1 | XP_011531617.1 | 4 - 5 | 685 | 67 | |
XM_011533316.2 | XP_011531618.1 | 2 - 3 | 504 | 67 | |
GenBank mRNA | AF040703.2 | - | 2 - 3 | 313 | 67 |
AF061836.1 | - | 2 - 3 | 396 | 67 | |
AF102770.2 | - | 3 - 4 | 501 | 67 | |
AF102771.2 | - | 3 - 4 | 513 | 67 | |
AF102772.2 | - | 3 - 4 | 513 | 67 | |
AF132675.1 | - | 3 - 4 | 503 | 67 | |
AF132676.1 | - | 2 - 3 | 266 | 67 | |
AF132677.1 | - | 3 - 4 | 293 | 67 | |
AF286217.1 | - | 2 - 3 | 394 | 67 | |
AK292638.1 | - | 2 - 3 | 390 | 67 | |
BC110412.1 | - | 3 - 4 | 306 | 67 | |
BC117153.1 | - | 3 - 4 | 572 | 67 | |
BC143879.1 | - | 3 - 4 | 572 | 67 | |
BI915594.1 | - | 2 - 3 | 411 | 67 | |
BM562405.1 | - | 3 - 4 | 609 | 67 | |
BQ072763.1 | - | 3 - 4 | 304 | 67 | |
BT020047.1 | - | 2 - 3 | 255 | 67 | |
BT020048.1 | - | 2 - 3 | 255 | 67 |