The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286828.1 | NP_001273757.1 | 1 - 2 | 114 | 87 |
NM_198469.3 | NP_940871.2 | 1 - 2 | 114 | 87 | |
XM_005251877.4 | XP_005251934.1 | 1 - 2 | 110 | 87 | |
XM_006717034.3 | XP_006717097.1 | 1 - 2 | 162 | 87 | |
XM_011518457.2 | XP_011516759.1 | 1 - 2 | 110 | 87 | |
GenBank mRNA | AK128877.1 | - | 1 - 2 | 111 | 87 |
AK302081.1 | - | 1 - 2 | 88 | 87 | |
BC133690.1 | - | 1 - 2 | 105 | 87 | |
BC143440.1 | - | 1 - 2 | 74 | 87 | |
HY011325.1 | - | 1 - 2 | 115 | 87 |