Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOCD
Entrez Gene ID
93649
Gene Name
myocardin
Gene Aliases
MYCD
Chromosome Location
Chr.17: 12665890 - 12768949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146312.2 NP_001139784.1 11 - 12 2502 62
XM_005256864.1 XP_005256921.1 12 - 13 2546 62
XM_017025342.1 XP_016880831.1 10 - 11 2376 62
GenBank mRNA AK097821.1 - 8 - 9 2124 62
AK292885.1 - 11 - 12 2371 62
AY764180.1 - 11 - 12 2495 62
BC126307.1 - 11 - 12 2261 62
BC143391.1 - 11 - 12 2261 62

View Full Product Details