Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNRC6B
Entrez Gene ID
23112
Gene Name
trinucleotide repeat containing 6B
Gene Aliases
-
Chromosome Location
Chr.22: 40044817 - 40335808 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024843.1 NP_001020014.1 13 - 14 1599 60
NM_001162501.1 NP_001155973.1 12 - 13 3920 60
NM_015088.2 NP_055903.2 10 - 11 3590 60
GenBank mRNA AB029016.2 - 10 - 11 3573 60
AK294519.1 - 12 - 13 3911 60
AK304087.1 - 7 - 8 1075 60
BC028626.2 - 13 - 14 1532 60
BX647630.1 - 7 - 8 2792 60

View Full Product Details