Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GRM8
- Entrez Gene ID
-
2918
- Gene Name
- glutamate metabotropic receptor 8
- Gene Aliases
-
GLUR8, GPRC1H, MGLUR8, mGlu8
- Chromosome Location
-
Chr.7: 126438598 - 127253294 on Build GRCh38
View Full Product Details