The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286577.1 | NP_001273506.1 | 23 - 24 | 4834 | 93 |
NM_015531.5 | NP_056346.3 | 23 - 24 | 4834 | 93 | |
XM_011544904.1 | XP_011543206.1 | 23 - 24 | 4733 | 93 | |
XM_011544905.1 | XP_011543207.1 | 23 - 24 | 4733 | 93 | |
XM_017017510.1 | XP_016872999.1 | 23 - 24 | 4733 | 93 | |
XM_017017511.1 | XP_016873000.1 | 22 - 23 | 4659 | 93 | |
XM_017017512.1 | XP_016873001.1 | 22 - 23 | 4624 | 93 | |
XM_017017513.1 | XP_016873002.1 | 13 - 14 | 2830 | 93 | |
XM_017017514.1 | XP_016873003.1 | 23 - 24 | 4733 | 93 | |
XM_017017515.1 | XP_016873004.1 | 9 - 10 | 2383 | 93 | |
XM_017017516.1 | XP_016873005.1 | 9 - 10 | 1953 | 93 | |
GenBank mRNA | AB231764.1 | - | 23 - 24 | 4648 | 93 |
AK091397.1 | - | 7 - 8 | 1352 | 93 | |
AK125894.1 | - | 10 - 11 | 2495 | 93 | |
AL833903.1 | - | 4 - 5 | 1031 | 93 | |
BC035599.1 | - | 3 - 4 | 885 | 93 | |
BC110508.2 | - | 10 - 11 | 2463 | 93 |