The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001315501.1 | NP_001302430.1 | 9 - 10 | 1395 | 62 |
NM_001315502.1 | NP_001302431.1 | 7 - 8 | 885 | 62 | |
NM_002555.5 | NP_002546.3 | 9 - 10 | 1179 | 62 | |
NM_183233.2 | NP_899056.2 | 9 - 10 | 1182 | 62 | |
GenBank mRNA | AF028738.1 | - | 9 - 10 | 1158 | 62 |
AF030302.1 | - | 9 - 10 | 1395 | 62 | |
AF037064.1 | - | 9 - 10 | 1166 | 62 | |
AF059663.1 | - | 9 - 10 | 1158 | 62 | |
AF070479.1 | - | 9 - 10 | 1158 | 62 | |
BC015571.2 | - | 9 - 10 | 1182 | 62 | |
BG684685.1 | - | 1 - 2 | 100 | 62 |