The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169122.1 | NP_001162593.1 | 13 - 14 | 3576 | 83 |
NM_001169123.1 | NP_001162594.1 | 14 - 15 | 3651 | 83 | |
NM_001169124.1 | NP_001162595.1 | 13 - 14 | 3576 | 83 | |
NM_001169125.1 | NP_001162596.1 | 13 - 14 | 3564 | 83 | |
NM_001170628.1 | NP_001164099.1 | 11 - 12 | 2215 | 83 | |
NM_002025.3 | NP_002016.2 | 14 - 15 | 3681 | 83 | |
GenBank mRNA | AB102644.1 | - | 13 - 14 | 3085 | 83 |
AJ001550.1 | - | 13 - 14 | 3085 | 83 | |
AK301927.1 | - | 11 - 12 | 2215 | 83 | |
BC132683.1 | - | 14 - 15 | 3223 | 83 | |
BC143740.1 | - | 13 - 14 | 3136 | 83 | |
BC143744.1 | - | 13 - 14 | 3148 | 83 | |
L76569.1 | - | 13 - 14 | 3489 | 83 | |
U48436.1 | - | 14 - 15 | 3681 | 83 |