The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004134.6 | NP_004125.3 | 12 - 13 | 1827 | 139 |
GenBank mRNA | AK222758.1 | - | 12 - 13 | 1549 | 139 |
AK225488.1 | - | 9 - 10 | 1174 | 139 | |
AK297795.1 | - | 13 - 14 | 1479 | 139 | |
AK297897.1 | - | 12 - 13 | 1528 | 139 | |
AK315177.1 | - | 12 - 13 | 1615 | 139 | |
BC000478.2 | - | 12 - 13 | 1558 | 139 | |
BC024034.2 | - | 12 - 13 | 1558 | 139 | |
BC030634.1 | - | 12 - 13 | 1522 | 139 | |
DQ480334.1 | - | 3 - 4 | 517 | 139 | |
L11066.1 | - | 12 - 13 | 1606 | 139 |