The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006725.4 | NP_006716.3 | 11 - 12 | 2066 | 111 |
XM_006718738.1 | XP_006718801.1 | 10 - 11 | 1970 | 111 | |
XM_006718739.1 | XP_006718802.1 | 10 - 11 | 1943 | 111 | |
XM_006718741.1 | XP_006718804.1 | 9 - 10 | 1847 | 111 | |
XM_011545362.1 | XP_011543664.1 | 10 - 11 | 1763 | 111 | |
GenBank mRNA | AK126159.1 | - | 2 - 3 | 1624 | 111 |
AK128419.1 | - | 12 - 13 | 1970 | 111 | |
AK292704.1 | - | 11 - 12 | 1942 | 111 | |
AK312749.1 | - | 11 - 12 | 2027 | 111 | |
BC078669.1 | - | 11 - 12 | 2005 | 111 | |
U66142.1 | - | 11 - 12 | 1841 | 111 |