The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018113.2 | NP_001018123.1 | 1 - 2 | 81 | 109 |
NR_136707.1 | 1 - 2 | 81 | 109 | ||
XM_011545470.2 | XP_011543772.1 | 1 - 2 | 267 | 109 | |
XM_017029356.1 | XP_016884845.1 | 1 - 2 | 267 | 109 | |
GenBank mRNA | BC043596.1 | - | 1 - 2 | 64 | 109 |
BC055411.1 | - | 1 - 2 | 74 | 109 | |
BC136558.1 | - | 1 - 2 | 63 | 109 | |
BP236844.1 | - | 1 - 2 | 74 | 109 |