Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A11
Entrez Gene ID
55867
Gene Name
solute carrier family 22 member 11
Gene Aliases
OAT4, hOAT4
Chromosome Location
Chr.11: 64555601 - 64572875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018484.3 NP_060954.1 8 - 9 1785 68
XM_011545167.1 XP_011543469.1 7 - 8 1435 68
GenBank mRNA AB026116.1 - 8 - 9 1438 68
AK075224.1 - 8 - 9 1131 68
AK222869.1 - 8 - 9 1443 68
AK290791.1 - 8 - 9 1768 68
BC034384.1 - 8 - 9 1760 68

View Full Product Details