Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A28
Entrez Gene ID
81894
Gene Name
solute carrier family 25 member 28
Gene Aliases
MFRN2, MRS3/4, MRS4L, NPD016
Chromosome Location
Chr.10: 99610518 - 99659329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005270208.1 XP_005270265.1 1 - 2 417 69

View Full Product Details