The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031212.3 | NP_112489.3 | 2 - 3 | 650 | 120 |
XM_005270208.1 | XP_005270265.1 | 2 - 3 | 640 | 120 | |
XM_005270211.4 | XP_005270268.1 | 2 - 3 | 254 | 120 | |
XM_006718005.3 | XP_006718068.1 | 2 - 3 | 314 | 120 | |
XM_006718006.3 | XP_006718069.1 | 2 - 3 | 645 | 120 | |
XM_011540239.2 | XP_011538541.1 | 2 - 3 | 536 | 120 | |
XM_011540240.1 | XP_011538542.1 | 3 - 4 | 1786 | 120 | |
XM_011540244.2 | XP_011538546.1 | 2 - 3 | 645 | 120 | |
XM_017016741.1 | XP_016872230.1 | 2 - 3 | 1722 | 120 | |
GenBank mRNA | AF327402.1 | - | 2 - 3 | 550 | 120 |
AJ303077.1 | - | 2 - 3 | 550 | 120 | |
BC047312.1 | - | 2 - 3 | 597 | 120 | |
BC058937.1 | - | 2 - 3 | 559 | 120 | |
BC076399.1 | - | 2 - 3 | 590 | 120 | |
BC094821.1 | - | 2 - 3 | 333 | 120 |