The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014146.3 | NP_054865.2 | 7 - 8 | 666 | 92 |
NM_032463.2 | NP_115852.1 | 8 - 9 | 785 | 92 | |
NM_032464.2 | NP_115853.2 | 8 - 9 | 855 | 92 | |
XM_011516558.2 | XP_011514860.1 | 8 - 9 | 695 | 92 | |
GenBank mRNA | AF252611.1 | - | 8 - 9 | 606 | 92 |
AF252613.1 | - | 7 - 8 | 402 | 92 | |
AF252614.1 | - | 8 - 9 | 536 | 92 | |
AF257135.1 | - | 6 - 7 | 337 | 92 | |
AK002099.1 | - | 8 - 9 | 645 | 92 | |
AK092904.1 | - | 8 - 9 | 996 | 92 | |
AK225150.1 | - | 7 - 8 | 417 | 92 | |
AK290074.1 | - | 8 - 9 | 533 | 92 | |
AK290916.1 | - | 7 - 8 | 414 | 92 | |
AY190023.1 | - | 6 - 7 | 347 | 92 | |
BC001609.1 | - | 8 - 9 | 667 | 92 | |
BC009204.2 | - | 6 - 7 | 339 | 92 |