The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002914.4 | NP_002905.2 | 3 - 4 | 299 | 78 |
NM_181471.2 | NP_852136.1 | 3 - 4 | 299 | 78 | |
XM_017012492.1 | XP_016867981.1 | 3 - 4 | 277 | 78 | |
GenBank mRNA | AB451243.1 | - | 3 - 4 | 224 | 78 |
AB451368.1 | - | 3 - 4 | 224 | 78 | |
BC002813.1 | - | 3 - 4 | 229 | 78 | |
BQ070526.1 | - | 3 - 4 | 249 | 78 | |
BT007058.1 | - | 3 - 4 | 224 | 78 | |
HY046046.1 | - | 3 - 4 | 300 | 78 | |
HY056452.1 | - | 3 - 4 | 293 | 78 | |
M87338.1 | - | - | 431 | 78 |