The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278791.1 | NP_001265720.1 | 6 - 7 | 714 | 59 |
NM_001278792.1 | NP_001265721.1 | 5 - 6 | 505 | 59 | |
NM_001278793.1 | NP_001265722.1 | 5 - 6 | 612 | 59 | |
NM_002914.4 | NP_002905.2 | 5 - 6 | 510 | 59 | |
NM_181471.2 | NP_852136.1 | 6 - 7 | 612 | 59 | |
XM_006716080.2 | XP_006716143.1 | 4 - 5 | 334 | 59 | |
XM_017012492.1 | XP_016867981.1 | 6 - 7 | 590 | 59 | |
XM_017012493.1 | XP_016867982.1 | 4 - 5 | 358 | 59 | |
GenBank mRNA | AB451243.1 | - | 6 - 7 | 537 | 59 |
AB451368.1 | - | 6 - 7 | 537 | 59 | |
BC002813.1 | - | 5 - 6 | 440 | 59 | |
BM563468.1 | - | 6 - 7 | 643 | 59 | |
BM810551.1 | - | 5 - 6 | 417 | 59 | |
BQ070526.1 | - | 6 - 7 | 562 | 59 | |
BQ674131.1 | - | 5 - 6 | 463 | 59 | |
BT007058.1 | - | 5 - 6 | 435 | 59 | |
M87338.1 | - | - | 744 | 59 |