The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129994.1 | NP_001123466.1 | 6 - 7 | 968 | 59 |
NM_001129995.1 | NP_001123467.1 | 6 - 7 | 965 | 59 | |
XM_011527296.1 | XP_011525598.1 | 6 - 7 | 951 | 59 | |
XM_011527297.2 | XP_011525599.1 | 7 - 8 | 1002 | 59 | |
XM_011527298.2 | XP_011525600.1 | 7 - 8 | 1037 | 59 | |
XM_017027283.1 | XP_016882772.1 | 7 - 8 | 1093 | 59 | |
GenBank mRNA | AK027901.1 | - | 6 - 7 | 961 | 59 |
AK096843.1 | - | 5 - 6 | 990 | 59 | |
AK225793.1 | - | 6 - 7 | 933 | 59 | |
AK291465.1 | - | 6 - 7 | 953 | 59 | |
AK294543.1 | - | 7 - 8 | 1037 | 59 |