Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLLT10
Entrez Gene ID
8028
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10
Gene Aliases
AF10
Chromosome Location
Chr.10: 21533349 - 21743630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004641.3 NP_004632.1 12 - 13 2010 73
NR_136736.1 13 - 14 2188 73
GenBank mRNA AB209755.1 - 13 - 14 1894 73
U13948.1 - 12 - 13 1844 73

View Full Product Details